COM448685661 Combi-Blocks, Inc.

3-(3,5-Dichlorophenyl)-5-methoxybenzoic acid

  • MDL number MFCD18321923
  • Cas Number : 1261954-74-8
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2226

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2226
Availability details for
Quantity
  • Availability
  • Add to Cart