COM448685662 Combi-Blocks, Inc.

5-Chloro-3-(3,5-dichlorophenyl)benzoic acid

  • MDL number MFCD18321927
  • Cas Number : 1261914-46-8
  • Molecular Weight : 301.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2230
Availability details for
Quantity
  • Availability
  • Add to Cart