COM448685663 Combi-Blocks, Inc.

3-[3-(N,N-Dimethylaminocarbonyl)phenyl]-5-methoxybenzoic acid

  • MDL number MFCD18321954
  • Cas Number : 1261907-71-4
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2257
Availability details for
Quantity
  • Availability
  • Add to Cart