COM448685664 Combi-Blocks, Inc.

4-[4-(N,N-Dimethylaminocarbonyl)phenyl]-3-methylbenzoic acid

  • MDL number MFCD18321967
  • Cas Number : 1261915-37-0
  • Molecular Weight : 283.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2270
Availability details for
Quantity
  • Availability
  • Add to Cart