COM448685669 Combi-Blocks, Inc.

2-Chloro-4-[3-(cyclopropylaminocarbonyl)phenyl]benzoic acid

  • MDL number MFCD18322387
  • Cas Number : 1261984-00-2
  • Molecular Weight : 315.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2694
Availability details for
Quantity
  • Availability
  • Add to Cart