COM448685682 Combi-Blocks, Inc.

5-(3,5-Dicarboxyphenyl)-2-fluorobenzoic acid

  • MDL number MFCD18322532
  • Cas Number : 1261984-63-7
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2840
Availability details for
Quantity
  • Availability
  • Add to Cart