COM448685686 Combi-Blocks, Inc.

4-(3-Benzyloxyphenyl)-3-methylbenzoic acid

  • MDL number MFCD18322860
  • Cas Number : 1261911-97-0
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3169
Availability details for
Quantity
  • Availability
  • Add to Cart