COM448685689 Combi-Blocks, Inc.

4-(3-Benzyloxyphenyl)-2-chlorobenzoic acid

  • MDL number MFCD18322880
  • Cas Number : 1261938-67-3
  • Molecular Weight : 338.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3190
Availability details for
Quantity
  • Availability
  • Add to Cart