COM448685691 Combi-Blocks, Inc.

4-(4-Benzyloxyphenyl)-3-methylbenzoic acid

  • MDL number MFCD18322890
  • Cas Number : 167627-35-2
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3200

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3200
Availability details for
Quantity
  • Availability
  • Add to Cart