COM448685694 Combi-Blocks, Inc.

4-(4-Benzyloxyphenyl)-2-methoxybenzoic acid

  • MDL number MFCD18322909
  • Cas Number : 1261913-24-9
  • Molecular Weight : 334.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3219

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3219
Availability details for
Quantity
  • Availability
  • Add to Cart