COM448685696 Combi-Blocks, Inc.

3-(2-N,N-Dimethylsulfamoylphenyl)-4-fluorobenzoic acid

  • MDL number MFCD18322930
  • Cas Number : 1261899-25-5
  • Molecular Weight : 323.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3240

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3240
Availability details for
Quantity
  • Availability
  • Add to Cart