COM448685697 Combi-Blocks, Inc.

3-(2-N,N-Dimethylsulfamoylphenyl)-5-fluorobenzoic acid

  • MDL number MFCD18322934
  • Cas Number : 1261996-26-2
  • Molecular Weight : 323.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3244

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3244
Availability details for
Quantity
  • Availability
  • Add to Cart