COM448685699 Combi-Blocks, Inc.

5-Methoxy-3-[4-(piperidinocarbonyl)phenyl]benzoic acid

  • MDL number MFCD18323032
  • Cas Number : 1261969-11-2
  • Molecular Weight : 339.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3342

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3342
Availability details for
Quantity
  • Availability
  • Add to Cart