COM448685701 Combi-Blocks, Inc.

4-Phenylnicotinic acid

  • MDL number MFCD04114252
  • Cas Number : 103863-14-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3602
Availability details for
Quantity
  • Availability
  • Add to Cart