COM448685705 Combi-Blocks, Inc.

5-Hydroxy-2-(thiophen-2-yl)pyridine

  • MDL number MFCD12033398
  • Cas Number : 1159815-70-9
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3613
Availability details for
Quantity
  • Availability
  • Add to Cart