COM448685708 Combi-Blocks, Inc.

4-(4-Fluorophenyl)nicotinic acid

  • MDL number MFCD18207551
  • Cas Number : 32923-72-1
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3654
Availability details for
Quantity
  • Availability
  • Add to Cart