COM448685712 Combi-Blocks, Inc.

2-(3-Formylphenyl)-4-hydroxypyridine

  • MDL number MFCD18323337
  • Cas Number : 1261909-98-1
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3676
Availability details for
Quantity
  • Availability
  • Add to Cart