COM448685713 Combi-Blocks, Inc.

5-Hydroxy-2-(2-methoxyphenyl)pyridine

  • MDL number MFCD17677688
  • Cas Number : 1255638-36-8
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3717

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3717
Availability details for
Quantity
  • Availability
  • Add to Cart