COM448685714 Combi-Blocks, Inc.

5-Hydroxy-2-(4-methoxyphenyl)pyridine

  • MDL number MFCD17677747
  • Cas Number : 1255636-82-8
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3725
Availability details for
Quantity
  • Availability
  • Add to Cart