COM448685715 Combi-Blocks, Inc.

2-(2,5-Difluorophenyl)-5-hydroxypyridine

  • MDL number MFCD18323451
  • Cas Number : 1261937-03-4
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3809
Availability details for
Quantity
  • Availability
  • Add to Cart