COM448685717 Combi-Blocks, Inc.

2-[Benzo(b)thiophen-2-yl]-5-hydroxypyridine

  • MDL number MFCD18323585
  • Cas Number : 1261998-84-8
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3949

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3949
Availability details for
Quantity
  • Availability
  • Add to Cart