COM448685718 Combi-Blocks, Inc.

6-Amino-3-(4-t-butylphenyl)picolinic acid

  • MDL number MFCD18323587
  • Cas Number : 1261913-25-0
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-3951
Availability details for
Quantity
  • Availability
  • Add to Cart