COM448685720 Combi-Blocks, Inc.

5-Hydroxy-2-(4-methylsulfonylphenyl)pyridine

  • MDL number MFCD18323740
  • Cas Number : 1032825-20-9
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4113
Availability details for
Quantity
  • Availability
  • Add to Cart