COM448685723 Combi-Blocks, Inc.

Methyl 3-methoxybiphenyl-4-carboxylate

  • MDL number MFCD06204218
  • Cas Number : 175152-70-2
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4243
Availability details for
Quantity
  • Availability
  • Add to Cart