COM448685726 Combi-Blocks, Inc.

Methyl 2-(4-formylphenyl)benzoate

  • MDL number MFCD06801752
  • Cas Number : 144291-47-4
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4249
Availability details for
Quantity
  • Availability
  • Add to Cart