COM448685731 Combi-Blocks, Inc.

4-Bromo-3'-fluorobiphenyl

  • MDL number MFCD06201408
  • Cas Number : 40641-65-4
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4256
Availability details for
Quantity
  • Availability
  • Add to Cart