COM448685734 Combi-Blocks, Inc.

4-(4-Methylphenyl)aniline

  • MDL number MFCD01740420
  • Cas Number : 1204-78-0
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4264
Availability details for
Quantity
  • Availability
  • Add to Cart