COM448685736 Combi-Blocks, Inc.

1-(4-methoxyphenyl)-4-nitrobenzene

  • MDL number MFCD00143334
  • Cas Number : 2143-90-0
  • Molecular Weight : 229.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4266
Availability details for
Quantity
  • Availability
  • Add to Cart