COM448685744 Combi-Blocks, Inc.

5-(2-Chloro-4-methylphenyl)pyridin-2-amine

  • MDL number MFCD18084567
  • Cas Number : 1258634-27-3
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4287

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4287
Availability details for
Quantity
  • Availability
  • Add to Cart