COM448685747 Combi-Blocks, Inc.

3-(6-Aminopyridin-3-yl)-N-ethylbenzamide

  • MDL number MFCD19684155
  • Cas Number : 1314987-59-1
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4298

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4298
Availability details for
Quantity
  • Availability
  • Add to Cart