COM448685749 Combi-Blocks, Inc.

Ethyl 4-(2-aminopyridin-5-yl)benzoate

  • MDL number MFCD19684157
  • Cas Number : 1314987-69-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4300

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4300
Availability details for
Quantity
  • Availability
  • Add to Cart