COM448685750 Combi-Blocks, Inc.

3-(6-Aminopyridin-3-yl)-N,N-dimethylbenzamide

  • MDL number MFCD19684163
  • Cas Number : 1314988-25-4
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4306

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4306
Availability details for
Quantity
  • Availability
  • Add to Cart