COM448685751 Combi-Blocks, Inc.

4-(6-Aminopyridin-3-yl)-N,N-dimethylbenzamide

  • MDL number MFCD19684164
  • Cas Number : 1314987-53-5
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4307
Availability details for
Quantity
  • Availability
  • Add to Cart