COM448685753 Combi-Blocks, Inc.

Methyl 5-(6-aminopyridin-3-yl)-2-fluorobenzoate

  • MDL number MFCD19684172
  • Cas Number : 1314988-17-4
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4315

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4315
Availability details for
Quantity
  • Availability
  • Add to Cart