COM448685756 Combi-Blocks, Inc.

Methyl 3-(6-aminopyridin-3-yl)benzoate

  • MDL number MFCD06802150
  • Cas Number : 889950-25-8
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4322

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4322
Availability details for
Quantity
  • Availability
  • Add to Cart