COM448685758 Combi-Blocks, Inc.

2-Amino-5-(4-methoxyphenyl)pyridine

  • MDL number MFCD06802137
  • Cas Number : 503536-75-2
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4341
Availability details for
Quantity
  • Availability
  • Add to Cart