COM448685759 Combi-Blocks, Inc.

5-(3-Methoxyphenyl)pyridin-2-amine

  • MDL number MFCD06802136
  • Cas Number : 893738-18-6
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4343
Availability details for
Quantity
  • Availability
  • Add to Cart