COM448685763 Combi-Blocks, Inc.

2-Amino-5-[4-fluoro-3-(trifluoromethyl)phenyl]pyridine

  • MDL number MFCD19684196
  • Cas Number : 1110656-35-3
  • Molecular Weight : 256.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4372
Availability details for
Quantity
  • Availability
  • Add to Cart