COM448685764 Combi-Blocks, Inc.

3-(6-Aminopyridin-3-yl)benzoic acid

  • MDL number MFCD06802729
  • Cas Number : 779315-67-2
  • Molecular Weight : 214.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4377

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4377
Availability details for
Quantity
  • Availability
  • Add to Cart