COM448685773 Combi-Blocks, Inc.

2-Amino-5-[4-(piperidinocarbonyl)phenyl]pyridine

  • MDL number MFCD19684208
  • Cas Number : 1314987-03-5
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4396

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4396
Availability details for
Quantity
  • Availability
  • Add to Cart