COM448685778 Combi-Blocks, Inc.

5-[3-(pyrrolidinocarbonyl]phenyl]pyridin-2-amine

  • MDL number MFCD19684213
  • Cas Number : 1314988-21-0
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4406

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4406
Availability details for
Quantity
  • Availability
  • Add to Cart