COM448685784 Combi-Blocks, Inc.

4-Benzyloxy-2'-(trifluoromethyl)biphenyl

  • MDL number MFCD19684220
  • Cas Number : 1314988-09-4
  • Molecular Weight : 328.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4426
Availability details for
Quantity
  • Availability
  • Add to Cart