COM448685788 Combi-Blocks, Inc.

Methyl 2-(2-methoxyphenyl)benzoate

  • MDL number MFCD06204198
  • Cas Number : 63506-58-1
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4432
Availability details for
Quantity
  • Availability
  • Add to Cart