COM448685790 Combi-Blocks, Inc.

Ethyl 6-(2,3-difluorophenyl)picolinate

  • MDL number MFCD18405135
  • Cas Number : 1261831-63-3
  • Molecular Weight : 263.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4434

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4434
Availability details for
Quantity
  • Availability
  • Add to Cart