COM448685792 Combi-Blocks, Inc.

Methyl 2-(3-chlorophenyl)benzoate

  • MDL number MFCD05979624
  • Cas Number : 773134-22-8
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4436
Availability details for
Quantity
  • Availability
  • Add to Cart