COM448685795 Combi-Blocks, Inc.

4-(2-Cyanophenyl)benzoic acid

  • MDL number MFCD06802562
  • Cas Number : 5728-44-9
  • Molecular Weight : 223.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4439
Availability details for
Quantity
  • Availability
  • Add to Cart