COM448685812 Combi-Blocks, Inc.

4-(3,5-Difluorophenyl)phenylacetic acid

  • MDL number MFCD03839974
  • Cas Number : 886363-26-4
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4456

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4456
Availability details for
Quantity
  • Availability
  • Add to Cart