COM448685813 Combi-Blocks, Inc.

4-(3-Cyanophenyl)phenylacetic acid

  • MDL number MFCD05980727
  • Cas Number : 893640-23-8
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4457
Availability details for
Quantity
  • Availability
  • Add to Cart