COM448685816 Combi-Blocks, Inc.

4-[4-(Dimethylcarbamoyl)phenyl]phenylacetic acid

  • MDL number MFCD20231501
  • Cas Number : 1125447-12-2
  • Molecular Weight : 283.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4461

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4461
Availability details for
Quantity
  • Availability
  • Add to Cart