COM448685821 Combi-Blocks, Inc.

Methyl 3-(4-aminophenyl)-5-fluorobenzoate

  • MDL number MFCD20231504
  • Cas Number : 1334499-98-7
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-4466
Availability details for
Quantity
  • Availability
  • Add to Cart